• USA Home
  • Search Results

  • cis3hexenylacetate14220368171811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-3-Hexenyl acetate

3 Products
Synonym:   (3Z)-3-Hexen-1-ol acetate, (3Z)-C-3-Hexenyl acetate, Leaf acetate
Image of <I>cis</I>-3-Hexenyl acetate
CAS Number:
Linear Formula:
 CH3CO2CH2CH2CH=CHC2H5
Molecular Weight:
 142.20
 Council of Europe no.:
 644
 EC Number:
 FEMA Number:
 3171
 Flavis number:
 9.197
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Document Links
  Application(s)
  Allergen
  Organoleptic property
  Assay
  Special Grade
Show More