• USA Home
  • Search Results

  • cis3hexenylhexanoate198303150111811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-3-Hexenyl hexanoate

2 Products
Image of <I>cis</I>-3-Hexenyl hexanoate
CAS Number:
Linear Formula:
 CH3(CH2)4CO2CH2CH2CH=CHC2H5
Molecular Weight:
 198.30
 FEMA Number:
 3403
 Flavis number:
 9.271
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Agency/Method
Show More