• USA Home
  • Search Results

  • cis4decenal154252166209911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-4-Decenal

2 Products
Image of <I>cis</I>-4-Decenal
CAS Number:
Linear Formula:
 CH3(CH2)4CH=CHCH2CH2CHO
Molecular Weight:
 154.25
 Council of Europe no.:
 2297
 EC Number:
 FEMA Number:
 3264
 Flavis number:
 5.137
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More