• USA Home
  • Search Results

  • cis4hydroxylproline1311361827911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-4-Hydroxy-L-proline

1 Products
Synonym:   (2S,4S)-(−)-4-Hydroxy-2-pyrrolidinecarboxylic acid, CHP
Image of <I>cis</I>-4-Hydroxy-<SC>L</SC>-proline
CAS Number:
Empirical Formula (Hill Notation):
 C5H9NO3
Molecular Weight:
 131.13
 Beilstein/REAXYS Number:
 81440
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More