• USA Home
  • Search Results

  • cis58111417eicosapentaenoicacid302451041794411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-5,8,11,14,17-Eicosapentaenoic acid

4 Products
Synonym:   EPA, Timnodonic acid
Image of <I>cis</I>-<WBR>5,8,11,14,17-<WBR>Eicosapentaenoic acid
CAS Number:
Linear Formula:
 CH3(CH2CH=CH)5(CH2)3CO2H
Molecular Weight:
 302.45
 Beilstein/REAXYS Number:
 1714433
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Titertest
  Assay
Show More