• USA Home
  • Search Results

  • cis81114eicosatrienoicacidmethylester320512106110911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-8,11,14-Eicosatrienoic acid methyl ester

1 Products
Synonym:   Dihomo-γ-linolenic acid methyl ester, Fame 20:3n-6, Methyl DGLA
Image of <I>cis</I>-<WBR>8,11,14-<WBR>Eicosatrienoic acid methyl ester
CAS Number:
Empirical Formula (Hill Notation):
 C21H36O2
Molecular Weight:
 320.51
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More