• USA Home
  • Search Results

  • cisdiamineplatinumiidichloride300051566327111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-Diamineplatinum(II) dichloride

1 Products
Synonym:   cis-Diammineplatinum(II) dichloride, cis-Dichlorodiammine platinum(II), cis-Platinum(II) diammine dichloride, Cisplatin
Image of <I>cis</I>-Diamineplatinum(II) dichloride
CAS Number:
Linear Formula:
 Pt(NH3)2Cl2
Molecular Weight:
 300.05
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More