• USA Home
  • Search Results

  • cisstilbene1802564549811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-Stilbene

1 Products
Synonym:   cis-1,2-Diphenylethylene, Isostilbene
Image of <I>cis</I>-Stilbene
CAS Number:
Linear Formula:
 C6H5CH=CHC6H5
Molecular Weight:
 180.25
 Beilstein/REAXYS Number:
 1616739
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
S4808 cis-Stilbene, 96%,