• USA Home
  • Search Results

  • citraconicanhydride1120861602411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citraconic anhydride

2 Products
Synonym:   2-Methylmaleic anhydride, 3-Methyl-2,5-furandione, Citraconic acid anhydride, Methylmaleic anhydride, Monomethylmaleic anhydride
Image of Citraconic anhydride
CAS Number:
Empirical Formula (Hill Notation):
 C5H4O3
Molecular Weight:
 112.08
 Beilstein/REAXYS Number:
 1835
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
Show More