• USA Home
  • Search Results

  • citraldimethylacetal19830754937311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citral dimethyl acetal

1 Products
Synonym:   3,7-Dimethyl-2,6-octadienal dimethyl acetal
Image of Citral dimethyl acetal
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)=CHCH(OCH3)2
Molecular Weight:
 198.30
 Council of Europe no.:
 39
 EC Number:
 FEMA Number:
 2305
 Flavis number:
 6.005
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More