• USA Home
  • Search Results

  • citrateconcentratedsolution258076804211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citrate Concentrated Solution

2 Products
Image of Citrate Concentrated Solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H5Na3O7
Molecular Weight:
 258.07
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Product Line
Show More