• USA Home
  • Search Results

  • citricacid192127792911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citric acid

17 Products
Synonym:   2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid
Image of Citric acid
CAS Number:
Linear Formula:
 HOC(COOH)(CH2COOH)2
Molecular Weight:
 192.12
 Beilstein/REAXYS Number:
 782061
 E Number:
 E330
 EC Index Number:
 201-069-1
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
  Document Links
  Allergen
  Organoleptic property
  UV-Absorption Values
  Biological Source
  Special Grade
  Anal. Standard Form
  Suitability
  Vapor pressure
  Potency
  Density
  Bulk Density
  pH Value
  Application(s)
  Quality clarifier
  Physical Form
  Solubility
  Assay
  Agency/Method
Show More

Product #
Description
251275 Citric acid, ACS reagent, ≥99.5%,
C2404 Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture,
C0759 Citric acid, 99%,
C83155 Citric acid, 99%,
27109 Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance),
46933 Citric acid, analytical standard,
27487 Citric acid, BioUltra, anhydrous, ≥99.5% (T),
W230618 Citric acid, ≥99.5%, FCC, FG,
1.00241 Citric acid, anhydrous powder EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,E 330,FCC,
1.00247 Citric acid, anhydrous fine-granular EMPROVE® ESSENTIAL Ph Eur,BP,JP,USP,E 330,FCC,
1.37002 Citric acid, anhydrous powder EMPROVE® EXPERT Ph Eur,BP,JP,USP,ACS,
8.18707 Citric acid, anhydrous for synthesis,
791725 Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%,
1134368 Citric acid, United States Pharmacopeia (USP) Reference Standard,
A1202000 Citric acid, anhydrous, European Pharmacopoeia (EP) Reference Standard,
94676 Citric acid, certified reference material, TraceCERT®,
W230633 Citric acid, natural, ≥99.5%, FCC, FG,