• USA Home
  • Search Results

  • citronellaoiljava12345800029111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citronella oil, Java

2 Products
Synonym:   Citronella grass oil, Cymbopogon winterianus
CAS Number:
 FEMA Number:
 2308
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Geographic origin
  Agency/Method
Show More