• USA Home
  • Search Results

  • citropten2061948706911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Citropten

2 Products
Synonym:   5,7-Dimethoxycoumarin, Citropten, Limettin
Image of Citropten
CAS Number:
Empirical Formula (Hill Notation):
 C11H10O4
Molecular Weight:
 206.19
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More