• USA Home
  • Search Results

  • clenbuterolhydrochloride313652189819111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clenbuterol hydrochloride

3 Products
Synonym:   4-Amino-α-(t-butylaminomethyl)-3,5-dichlorobenzyl alcohol hydrochloride
Image of Clenbuterol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H18Cl2N2O · HCl
Molecular Weight:
 313.65
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
  Application(s)
Show More