• USA Home
  • Search Results

  • clindamycin2phosphate504962472996211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clindamycin 2-phosphate

1 Products
Synonym:   Clindamycin 2-dihydrogen phosphate
Image of Clindamycin 2-phosphate
CAS Number:
Empirical Formula (Hill Notation):
 C18H34ClN2O8PS
Molecular Weight:
 504.96
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More