• USA Home
  • Search Results

  • clindamycinphosphate504962472996211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clindamycin phosphate

3 Products
Synonym:   Clindamycin 2-dihydrogen phosphate, Clindamycin 2-phosphate
Image of Clindamycin phosphate
CAS Number:
Empirical Formula (Hill Notation):
 C18H34ClN2O8PS
Molecular Weight:
 504.96
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More