• USA Home
  • Search Results

  • clomipraminehydrochloride351311732177611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clomipramine hydrochloride

5 Products
Synonym:   3-Chloro-10,11-dihydro-N,N-dimethyl-5H-dibenz[b,f]azepine-5-propanamine hydrochloride, Anafranil hydrochloride
Image of Clomipramine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H23ClN2 · HCl
Molecular Weight:
 351.31
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Physical Form
  Special Grade
Show More