• USA Home
  • Search Results

  • clonazepam31571162261311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clonazepam

3 Products
Synonym:   5-(2-Chlorophenyl)-7-nitro-3H-1,4-benzodiazepin-2(1H)-one
Image of Clonazepam
CAS Number:
Empirical Formula (Hill Notation):
 C15H10ClN3O3
Molecular Weight:
 315.71
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More