• USA Home
  • Search Results

  • clonazepamimpurityb315715519889511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clonazepam impurity B

1 Products
Synonym:   3-Amino-4-(2-chlorophenyl)-6-nitro-2(1H)-quinolinone
Image of Clonazepam impurity B
CAS Number:
Empirical Formula (Hill Notation):
 C15H10ClN3O3
Molecular Weight:
 315.71
 Beilstein/REAXYS Number:
 5596577
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More