• USA Home
  • Search Results

  • clonazepamsolution31571162261311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clonazepam solution

1 Products
Image of Clonazepam solution
CAS Number:
Empirical Formula (Hill Notation):
 C15H10ClN3O3
Molecular Weight:
 315.71
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Anal. Standard Form
  Special Grade
  Titertest
Show More