• USA Home
  • Search Results

  • clonidinehydrochloride26655420591811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clonidine hydrochloride

4 Products
Synonym:   2-(2,6-Dichloroanilino)-2-imidazoline hydrochloride
Image of Clonidine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C9H9Cl2N3 · HCl
Molecular Weight:
 266.55
 Beilstein/REAXYS Number:
 4163525
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Solubility
  Special Grade
Show More