• USA Home
  • Search Results

  • cloprostenolsodiumsalthydrate44690anhydrousbasis55028723anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+/-)-Cloprostenol sodium salt hydrate

1 Products
Synonym:   (+/-)-16-(3-Chlorophenoxy)-9,11,15R-trihydroxy-17,18,19,20-tetranorprosta-5Z,13E-dien-1-oic acid sodium salt hydrate
Image of (+/-)-Cloprostenol sodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C22H28ClNaO6 · xH2O
Molecular Weight:
 446.90 (anhydrous basis)
 EC Number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More