• USA Home
  • Search Results

  • clotrimazole344842359375111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Clotrimazole

6 Products
Synonym:   1-(o-Chlorotrityl)imidazole, 1-(o-Chloro-α,α-diphenylbenzyl)imidazole, 1-[(2-Chlorophenyl)diphenylmethyl]-1H-imidazole
Image of Clotrimazole
CAS Number:
Empirical Formula (Hill Notation):
 C22H17ClN2
Molecular Weight:
 344.84
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Activity Spectrum
  Titertest
  Mode of Action
  Special Grade
  Anal. Standard Form
Show More