• USA Home
  • Search Results

  • cobalt5893744048411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cobalt

308 Products
Image of Cobalt
CAS Number:
Empirical Formula (Hill Notation):
 Co
Molecular Weight:
 58.93
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Purity Grade
  Weight
  Particle Dimension
  Composite Dimension
  Physical Form
  Assay
Show More

Product #
Description
697745 Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%,
266639 Cobalt, powder, 2 μm particle size, 99.8% trace metals basis,
266647 Cobalt, powder, <150 μm, ≥99.9% trace metals basis,
266655 Cobalt, pieces, 99.5% trace metals basis,
203076 Cobalt, granular, 99.99% trace metals basis,
356891 Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis,
266671 Cobalt, foil, thickness 0.25 mm, ≥99.99% trace metals basis,
356867 Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis,
266663 Cobalt, rod, diam. 5.0 mm, 99.998% trace metals basis,
398810 Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis,
266701 Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis,
326380 Cobalt, foil, thickness 0.1 mm, ≥99.99%,
GF94594450 Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.9%,
GF41541476 Cobalt, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF45372315 Cobalt, foil, 4mm disks, thickness 0.003mm, 99.9%,
GF08560289 Cobalt, foil, 4mm disks, thickness 0.004mm, 99.9%,
GF89559343 Cobalt, foil, 4mm disks, thickness 0.005mm, 99.9%,
GF48392051 Cobalt, foil, 4mm disks, thickness 0.006mm, 99.9%,
GF55954873 Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%,
GF92926727 Cobalt, foil, 4mm disks, thickness 0.008mm, 99.9%,
GF70445246 Cobalt, foil, 4mm disks, thickness 0.009mm, 99.9%,
GF40089225 Cobalt, foil, 4mm disks, thickness 0.0125mm, 99.9%,
GF48813723 Cobalt, foil, 100x100mm, thickness 0.125mm, annealed, 99.9%,
GF10782121 Cobalt, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%,
GF40808516 Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.9%,
GF18093461 Cobalt, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%,
GF30160291 Cobalt, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%,
GF81966224 Cobalt, foil, 100x100mm, thickness 0.7mm, annealed, 99.9%,
GF56494187 Cobalt, foil, 100x100mm, thickness 1.0mm, as rolled, 99.9%,
GF76259593 Cobalt, foil, 100x100mm, thickness 2.0mm, as rolled, 99.9%,
GF91564904 Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%,
GF88288699 Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%,
GF30959411 Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%,
GF67617153 Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%,
GF85563535 Cobalt, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%,
GF09035647 Cobalt, foil, 25x25mm, thickness 0.7mm, annealed, 99.9%,
GF41834011 Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.9%,
GF34805268 Cobalt, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%,
GF40133472 Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, annealed, 99.9%,
GF03773640 Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%,
GF06754406 Cobalt, foil, light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%,
GF42159315 Cobalt, foil, light tested, 25x25mm, thickness 0.009mm, 99.9%,
GF75010564 Cobalt, foil, light tested, 25x25mm, thickness 0.0125mm, 99.9%,
GF53589897 Cobalt, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%,
GF15401135 Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%,
GF84130792 Cobalt, foil, light tested, 25x25mm, thickness 0.025mm, 99.9%,
GF04944539 Cobalt, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%,
GF36086250 Cobalt, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%,
GF69926462 Cobalt, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%,
GF52144747 Cobalt, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%,
GF29455724 Cobalt, foil, 50x50mm, thickness 0.7mm, annealed, 99.9%,
GF29251953 Cobalt, foil, 50x50mm, thickness 1.0mm, as rolled, 99.9%,
GF50658734 Cobalt, foil, 50x50mm, thickness 1.0mm, as rolled, 99.99+%,
GF00415426 Cobalt, foil, 10mm disks, thickness 0.003mm, 99.9%,
GF10413975 Cobalt, foil, 10mm disks, thickness 0.004mm, 99.9%,
GF89300022 Cobalt, foil, 10mm disks, thickness 0.005mm, 99.9%,
GF25878987 Cobalt, foil, 10mm disks, thickness 0.006mm, 99.9%,
GF18688731 Cobalt, foil, 10mm disks, thickness 0.008mm, 99.9%,
GF65365465 Cobalt, foil, 10mm disks, thickness 0.009mm, 99.9%,
GF59746168 Cobalt, foil, 10mm disks, thickness 0.0125mm, 99.9%,
GF87279924 Cobalt, foil, 10mm disks, thickness 0.015mm, 99.9%,
GF80385756 Cobalt, foil, 10mm disks, thickness 0.01mm, 99.9%,
GF50375886 Cobalt, foil, 10mm disks, thickness 0.02mm, 99.9%,
GF25719789 Cobalt, foil, 10mm disks, thickness 0.05mm, annealed, 99.9%,
GF44066864 Cobalt, foil, 10mm disks, thickness 0.05mm, as rolled, 99.9%,
GF10251582 Cobalt, foil, 10mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF39519164 Cobalt, foil, 10mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF41119657 Cobalt, foil, 10mm disks, thickness 0.25mm, as rolled, 99.9%,
GF56303456 Cobalt, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF51927565 Cobalt, foil, 15mm disks, thickness 0.003mm, 99.9%,
GF01747403 Cobalt, foil, 15mm disks, thickness 0.004mm, 99.9%,
GF62503220 Cobalt, foil, 15mm disks, thickness 0.005mm, 99.9%,
GF85070000 Cobalt, foil, 15mm disks, thickness 0.007mm, 99.9%,
GF36092011 Cobalt, foil, 15mm disks, thickness 0.008mm, 99.9%,
GF70682700 Cobalt, foil, 15mm disks, thickness 0.009mm, 99.9%,
GF68270362 Cobalt, foil, 15mm disks, thickness 0.0125mm, 99.9%,
GF09991154 Cobalt, foil, 15mm disks, thickness 0.015mm, 99.9%,
GF93200420 Cobalt, foil, 15mm disks, thickness 0.025mm, 99.9%,
GF96985591 Cobalt, foil, 15mm disks, thickness 0.02mm, 99.9%,
GF61689673 Cobalt, foil, 15mm disks, thickness 0.05mm, annealed, 99.9%,
GF08358116 Cobalt, foil, 15mm disks, thickness 0.05mm, as rolled, 99.9%,
GF81447554 Cobalt, foil, 15mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF39895273 Cobalt, foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF39956754 Cobalt, foil, 15mm disks, thickness 0.25mm, as rolled, 99.9%,
GF37932330 Cobalt, foil, 15mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF48894485 Cobalt, foil, 25mm disks, thickness 0.015mm, 99.9%,
GF27481732 Cobalt, foil, 25mm disks, thickness 0.01mm, 99.9%,
GF50052765 Cobalt, foil, 25mm disks, thickness 0.025mm, 99.9%,
GF55348140 Cobalt, foil, 25mm disks, thickness 0.05mm, annealed, 99.9%,
GF44764993 Cobalt, foil, 25mm disks, thickness 0.05mm, as rolled, 99.9%,
GF95811475 Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%,
GF62166629 Cobalt, microfoil, disks, 25mm, thinness 1.0μm, specific density 890μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF54906385 Cobalt, wire reel, 0.025m, diameter 2.0mm, hard, 99.99+%,
GF60120997 Cobalt, wire reel, 0.05m, diameter 0.5mm, hard, 99.99+%,
GF34640333 Cobalt, wire reel, 0.05m, diameter 1.0mm, hard, 99.99+%,
GF63298198 Cobalt, wire reel, 0.05m, diameter 2.0mm, hard, 99.99+%,
GF04914738 Cobalt, wire reel, 0.1m, diameter 0.125mm, hard, 99.99+%,
GF91382069 Cobalt, wire reel, 0.1m, diameter 0.1mm, hard, 99.99+%,
GF15124081 Cobalt, wire reel, 0.1m, diameter 0.5mm, hard, 99.99+%,
GF30041480 Cobalt, wire reel, 0.1m, diameter 1.0mm, hard, 99.99+%,
GF12274253 Cobalt, wire reel, 0.1m, diameter 2.0mm, hard, 99.99+%,
GF03600278 Cobalt, foil, 4mm disks, thickness 0.015mm, 99.9%,
GF78079630 Cobalt, foil, 4mm disks, thickness 0.01mm, 99.9%,
GF29762646 Cobalt, foil, 4mm disks, thickness 0.025mm, 99.9%,
GF97588300 Cobalt, foil, 4mm disks, thickness 0.02mm, 99.9%,
GF21186985 Cobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%,
GF76990337 Cobalt, foil, 4mm disks, thickness 0.05mm, as rolled, 99.9%,
GF69765890 Cobalt, foil, 4mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF95280396 Cobalt, foil, 4mm disks, thickness 0.125mm, annealed, 99.9%,
GF47765166 Cobalt, foil, 4mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF33091196 Cobalt, foil, 4mm disks, thickness 0.25mm, as rolled, 99.9%,
GF61683071 Cobalt, foil, 4mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF99449702 Cobalt, foil, 50mm disks, thickness 0.125mm, annealed, 99.9%,
GF71539229 Cobalt, foil, 6mm disks, thickness 0.003mm, 99.9%,
GF70035869 Cobalt, foil, 6mm disks, thickness 0.004mm, 99.9%,
GF94917718 Cobalt, foil, 6mm disks, thickness 0.005mm, 99.9%,
GF49006458 Cobalt, foil, 6mm disks, thickness 0.006mm, 99.9%,
GF94359853 Cobalt, foil, 6mm disks, thickness 0.007mm, 99.9%,
GF15841199 Cobalt, foil, 6mm disks, thickness 0.008mm, 99.9%,
GF05655382 Cobalt, foil, 6mm disks, thickness 0.009mm, 99.9%,
GF11152628 Cobalt, foil, 6mm disks, thickness 0.0125mm, 99.9%,
GF50034383 Cobalt, foil, 6mm disks, thickness 0.015mm, 99.9%,
GF59953944 Cobalt, foil, 6mm disks, thickness 0.01mm, 99.9%,
GF20330749 Cobalt, foil, 6mm disks, thickness 0.025mm, 99.9%,
GF98537620 Cobalt, foil, 6mm disks, thickness 0.02mm, 99.9%,
GF02500063 Cobalt, foil, 6mm disks, thickness 0.05mm, annealed, 99.9%,
GF23791620 Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.9%,
GF20736659 Cobalt, foil, 6mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF72500161 Cobalt, foil, 6mm disks, thickness 0.125mm, annealed, 99.9%,
GF70645721 Cobalt, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF36399437 Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.9%,
GF10384400 Cobalt, foil, 6mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF42403328 Cobalt, foil, 8mm disks, thickness 0.003mm, 99.9%,
GF54483318 Cobalt, foil, 8mm disks, thickness 0.004mm, 99.9%,
GF64714406 Cobalt, foil, 8mm disks, thickness 0.005mm, 99.9%,
GF25807035 Cobalt, foil, 8mm disks, thickness 0.006mm, 99.9%,
GF94981449 Cobalt, foil, 8mm disks, thickness 0.007mm, 99.9%,
GF80149507 Cobalt, foil, 8mm disks, thickness 0.008mm, 99.9%,
GF06922920 Cobalt, foil, 8mm disks, thickness 0.009mm, 99.9%,
GF94077962 Cobalt, foil, 8mm disks, thickness 0.0125mm, 99.9%,
GF54298233 Cobalt, foil, 8mm disks, thickness 0.015mm, 99.9%,
GF94243080 Cobalt, foil, 8mm disks, thickness 0.01mm, 99.9%,
GF41343566 Cobalt, foil, 8mm disks, thickness 0.025mm, 99.9%,
GF17203003 Cobalt, foil, 8mm disks, thickness 0.02mm, 99.9%,
GF84090742 Cobalt, foil, 8mm disks, thickness 0.05mm, annealed, 99.9%,
GF73952534 Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.9%,
GF92082012 Cobalt, foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%,
GF76991316 Cobalt, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%,
GF30716153 Cobalt, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%,
GF49928956 Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.9%,
GF98050428 Cobalt, foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%,
GF92700176 Cobalt, wire reel, 1m, diameter 0.1mm, hard, 99.99+%,
GF76579846 Cobalt, wire reel, 1m, diameter 0.25mm, hard, 99.99+%,
GF20239017 Cobalt, wire reel, 1m, diameter 0.5mm, hard, 99.99+%,
GF91340861 Cobalt, wire reel, 1m, diameter 0.05mm, hard, 99.9%,
GF58266478 Cobalt, wire reel, 2m, diameter 0.125mm, hard, 99.99+%,
GF87521542 Cobalt, wire reel, 2m, diameter 0.1mm, hard, 99.99+%,
GF72480966 Cobalt, wire reel, 2m, diameter 0.25mm, hard, 99.99+%,
GF17608091 Cobalt, wire reel, 2m, diameter 0.05mm, hard, 99.9%,
GF31361835 Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%,
GF76859892 Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%,
GF81964413 Cobalt, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%,
GF40621665 Cobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%,
GF13817516 Cobalt, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%,
GF09020078 Cobalt, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%,
GF72081592 Cobalt, foil, light tested, 50x50mm, thickness 0.025mm, 99.9%,
GF16350599 Cobalt, foil, light tested, 50x50mm, thickness 0.02mm, 99.9%,
GF46475220 Cobalt, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.9%,
GF20509354 Cobalt, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 99.99+%,
GF16330036 Cobalt, foil, not light tested, 100x100mm, thickness 0.003mm, 99.9%,
GF21348132 Cobalt, foil, not light tested, 100x100mm, thickness 0.004mm, 99.9%,
GF90540390 Cobalt, foil, not light tested, 100x100mm, thickness 0.005mm, 99.9%,
GF99715876 Cobalt, foil, not light tested, 100x100mm, thickness 0.006mm, 99.9%,
GF45144985 Cobalt, foil, not light tested, 100x100mm, thickness 0.007mm, 99.9%,
GF64339724 Cobalt, foil, not light tested, 100x100mm, thickness 0.008mm, 99.9%,
GF23494608 Cobalt, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%,
GF31168122 Cobalt, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.9%,
GF45956166 Cobalt, foil, not light tested, 100x100mm, thickness 0.01mm, 99.9%,
GF56754557 Cobalt, foil, not light tested, 100x100mm, thickness 0.025mm, 99.9%,
GF13124146 Cobalt, foil, not light tested, 100x100mm, thickness 0.02mm, 99.9%,
GF17839334 Cobalt, foil, not light tested, 100x100mm, thickness 0.05mm, as rolled, 99.9%,
GF64744937 Cobalt, foil, not light tested, 100x100mm, thickness 0.05mm, as rolled, 99.99+%,
GF44846208 Cobalt, foil, not light tested, 150x150mm, thickness 0.015mm, 99.9%,
GF13007180 Cobalt, foil, not light tested, 150x150mm, thickness 0.01mm, 99.9%,
GF03770995 Cobalt, foil, not light tested, 150x150mm, thickness 0.025mm, 99.9%,
GF77573650 Cobalt, foil, not light tested, 25x25mm, thickness 0.001mm, temporary acrylic support, 99.9%,
GF71622301 Cobalt, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.9%,
GF30616668 Cobalt, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.9%,
GF93295299 Cobalt, foil, not light tested, 25x25mm, thickness 0.004mm, 99.9%,
GF40673707 Cobalt, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%,
GF92296154 Cobalt, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%,
GF85339389 Cobalt, foil, not light tested, 25x25mm, thickness 0.007mm, 99.9%,
GF15928858 Cobalt, foil, not light tested, 25x25mm, thickness 0.008mm, 99.9%,
GF08123723 Cobalt, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%,
GF42387060 Cobalt, foil, not light tested, 50x50mm, thickness 0.001mm, temporary acrylic support, 99.9%,
GF37004464 Cobalt, foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.9%,
GF00561204 Cobalt, foil, not light tested, 50x50mm, thickness 0.002mm, temporary acrylic support, 99.9%,
GF17951160 Cobalt, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%,
GF76302839 Cobalt, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%,
GF79949738 Cobalt, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%,
GF73778776 Cobalt, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%,
GF79906382 Cobalt, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%,
GF31581200 Cobalt, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%,
GF00377029 Cobalt, foil, not light tested, 50x50mm, thickness 0.0125mm, 99.9%,
GF58271261 Cobalt, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%,
GF28651619 Cobalt, foil, not light tested, 50x50mm, thickness 0.025mm, 99.9%,
GF88007564 Cobalt, foil, not light tested, 50x50mm, thickness 0.02mm, 99.9%,
GF33428831 Cobalt, microfoil, disks, 10mm, thinness 0.1μm, specific density 94.6μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF41606131 Cobalt, microfoil, disks, 10mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF65219979 Cobalt, microfoil, disks, 10mm, thinness 1.0μm, specific density 890μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF25498187 Cobalt, microfoil, disks, 25mm, thinness 0.1μm, specific density 94.6μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF95862134 Cobalt, microfoil, disks, 25mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF78371747 Cobalt, microfoil, disks, 25mm, thinness 0.5μm, specific density 434μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF74432392 Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%,
GF14342026 Cobalt, wire reel, 0.2m, diameter 0.125mm, hard, 99.99+%,
GF22757935 Cobalt, wire reel, 0.2m, diameter 0.1mm, hard, 99.99+%,
GF70398502 Cobalt, wire reel, 0.2m, diameter 0.25mm, hard, 99.99+%,
GF90510094 Cobalt, wire reel, 0.2m, diameter 0.5mm, hard, 99.99+%,
GF55186050 Cobalt, wire reel, 0.2m, diameter 1.0mm, hard, 99.99+%,
GF34281480 Cobalt, wire reel, 0.2m, diameter 2.0mm, hard, 99.99+%,
GF64719579 Cobalt, wire reel, 0.2m, diameter 0.05mm, hard, 99.9%,
GF92752522 Cobalt, wire reel, 0.5m, diameter 0.125mm, hard, 99.99+%,
GF44741606 Cobalt, wire reel, 0.5m, diameter 0.1mm, hard, 99.99+%,
GF99812638 Cobalt, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%,
GF13761918 Cobalt, wire reel, 0.5m, diameter 0.5mm, hard, 99.99+%,
GF98256314 Cobalt, wire reel, 0.5m, diameter 1.0mm, hard, 99.99+%,
GF87130728 Cobalt, wire reel, 0.5m, diameter 0.05mm, hard, 99.9%,
GF94066139 Cobalt, wire reel, 1m, diameter 0.125mm, hard, 99.99+%,
GF95441283 Cobalt, foil, 50x50mm, thickness 0.5mm, as rolled, 99.99+%,
GF72680564 Cobalt, foil, 10mm disks, thickness 0.007mm, 99.9%,
GF67945607 Cobalt, foil, 10mm disks, thickness 0.025mm, 99.9%,
GF58641059 Cobalt, foil, 10mm disks, thickness 0.125mm, annealed, 99.9%,
GF34873320 Cobalt, foil, 15mm disks, thickness 0.006mm, 99.9%,
GF48219857 Cobalt, foil, 15mm disks, thickness 0.01mm, 99.9%,
GF79962137 Cobalt, foil, 15mm disks, thickness 0.125mm, annealed, 99.9%,
GF33795658 Cobalt, foil, 25mm disks, thickness 0.0125mm, 99.9%,
GF27157495 Cobalt, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%,
GF94203503 Cobalt, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%,
GF94218666 Cobalt, foil, not light tested, 25x25mm, thickness 0.009mm, 99.9%,
GF40290667 Cobalt, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%,
GF64168263 Cobalt, foil, not light tested, 50x50mm, thickness 0.015mm, 99.9%,
GF39774617 Cobalt, microfoil, disks, 10mm, thinness 0.5μm, specific density 434μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF14436503 Cobalt, wire reel, 0.05m, diameter 0.25mm, hard, 99.99+%,
GF04563187 Cobalt, wire reel, 0.1m, diameter 0.25mm, hard, 99.99+%,
GF01015300 Cobalt, powder, max. particle size 840 micron, weight 20 g, high purity 99.99+%,
GF01905308 Cobalt, lump, 40 mm max. lump size, weight 100 g, purity 99.8%,
GF02492995 Cobalt, rod, 2.0 mm diameter, length 2 mm, high purity 99.99+%,
GF05847880 Cobalt, lump, 40 mm max. lump size, weight 500 g, purity 99.8%,
GF05977558 Cobalt, rod, 6.35 mm diameter, length 25 mm, purity 99.9%,
GF06340597 Cobalt, powder, max. particle size 45 micron, weight 50 g, purity 99.8%,
GF09412455 Cobalt, foil, thickness 1.0 mm, size 250 × 250 mm, purity 99.9%,
GF13438751 Cobalt, powder, max. particle size 7.5 micron, weight 200 g, purity 99.6%,
GF14653807 Cobalt, rod, 6.35 mm diameter, length 50 mm, purity 99.9%,
GF14966231 Cobalt, powder, max. particle size 150 micron, weight 100 g, min. particle size 50 micron,
GF16866670 Cobalt, powder, max. particle size 150 micron, weight 200 g, min. particle size 50 micron,
GF17102764 Cobalt, rod, 2.0 mm diameter, length 200 mm, high purity 99.99+%,
GF18137729 Cobalt, rod, 3.0 mm diameter, length 50 mm, purity 99.9%,
GF21501795 Cobalt, powder, max. particle size 150 micron, weight 50 g, min. particle size 50 micron,
GF24195999 Cobalt, rod, 12 mm diameter, length 50 mm, purity 99.9%,
GF26913179 Cobalt, powder, max. particle size 840 micron, weight 50 g, high purity 99.99+%,
GF27025691 Cobalt, lump, 10 mm max. lump size, weight 100 g, high purity 99.99+%,
GF28351515 Cobalt, rod, 6.35 mm diameter, length 100 mm, purity 99.9%,
GF28563712 Cobalt, rod, 25 mm diameter, length 50 mm, purity 99.9%,
GF31725020 Cobalt, rod, 12 mm diameter, length 100 mm, purity 99.9%,
GF33483461 Cobalt, rod, 10 mm diameter, length 200 mm, purity 99.9%,
GF36809523 Cobalt, rod, 10 mm diameter, length 50 mm, purity 99.9%,
GF37773816 Cobalt, rod, 1.5 mm diameter, length 25 mm, high purity 99.99+%,
GF41793565 Cobalt, rod, 12 mm diameter, length 25 mm, purity 99.9%,
GF43939761 Cobalt, rod, 6.35 mm diameter, length 200 mm, purity 99.9%,
GF45491362 Cobalt, rod, 5.0 mm diameter, length 25 mm, high purity 99.99+%,
GF46166351 Cobalt, rod, 25 mm diameter, length 25 mm, purity 99.9%,
GF49028980 Cobalt, lump, 40 mm max. lump size, weight 200 g, purity 99.8%,
GF50744954 Cobalt, foil, thickness 2.0 mm, size 25 × 25 mm, purity 99.9%,
GF51020648 Cobalt, powder, mean particle size 0.02 micron, weight 20 g, purity 99.9%,
GF56174238 Cobalt, rod, 1.5 mm diameter, length 50 mm, high purity 99.99+%,
GF59680862 Cobalt, powder, 7.5 max. part. size (micron), weight 50 g, purity 99.6%,
GF60043317 Cobalt, lump, 40 mm max. lump size, weight 1000 g, purity 99.8%,
GF60107510 Cobalt, powder, 7.5 max. part. size (micron), weight 100 g, purity 99.6%,
GF63256743 Cobalt, powder, 150 max. part. size (micron), weight 500 g, min. particle size 50 micron,
GF64618544 Cobalt, rod, length 100 mm, 3.0 mm diameter, purity 99.9%,
GF65627679 Cobalt, rod, 12 mm diameter, length 200 mm, purity 99.9%,
GF67974328 Cobalt, lump, 10 mm max. lump size, weight 50 g, purity 99.9%,
GF68219174 Cobalt, rod, length 25 mm, 2.0 mm diameter, high purity 99.99+%,
GF69748644 Cobalt, rod, 25 mm diameter, length 100 mm, purity 99.9%,
GF70400405 Cobalt, foil, thickness 0.50 mm, size 50 × 50 mm, purity 99.9%,
GF71155469 Cobalt, rod, length 100 mm, 1.5 mm diameter, high purity 99.99+%,
GF71761451 Cobalt, rod, length 200 mm, 1.5 mm diameter, high purity 99.99+%,
GF71848503 Cobalt, powder, 45 max. part. size (micron), weight 200 g, purity 99.8%,
GF73396109 Cobalt, powder, 150 max. part. size (micron), weight 1000 g, min. particle size 50 micron,
GF75844356 Cobalt, lump, 10 mm max. lump size, weight 10 g, purity 99.9%,
GF76842984 Cobalt, rod, 2.0 mm diameter, length 50 mm, high purity 99.99+%,
GF77406756 Cobalt, powder, 0.02 mean particle size (micron), weight 10 g, purity 99.9%,
GF80270849 Cobalt, rod, 10 mm diameter, length 100 mm, purity 99.9%,
GF80543796 Cobalt, lump, 10 mm max. lump size, weight 20 g, purity 99.9%,
GF86373760 Cobalt, rod, 2.0 mm diameter, length 100 mm, high purity 99.99+%,
GF86421012 Cobalt, rod, 5.0 mm diameter, length 50 mm, high purity 99.99+%,
GF89168978 Cobalt, lump, 40 mm max. lump size, weight 2000 g, purity 99.8%,
GF89255290 Cobalt, lump, 10 mm max. lump size, weight 50 g, high purity 99.99+%,
GF90657164 Cobalt, foil, thickness 0.50 mm, size 125 × 125 mm, purity 99.9%,
GF92574782 Cobalt, powder, 45 max. part. size (micron), weight 100 g, purity 99.8%,
GF93598161 Cobalt, rod, 3.0 mm diameter, purity 99.9%, length 500 mm,
GF94023086 Cobalt, rod, 3.0 mm diameter, length 200 mm, purity 99.9%,
GF95107473 Cobalt, foil, thickness 1.0 mm, size 50 × 50 mm, purity 99.9%,
GF96008126 Cobalt, powder, 840 max. part. size (micron), weight 10 g, high purity 99.99+%,
GF96790113 Cobalt, lump, 10 mm max. lump size, weight 100 g, purity 99.9%,
GF97256559 Cobalt, lump, 10 mm max. lump size, weight 20 g, high purity 99.99+%,
GF97744742 Cobalt, rod, 10 mm diameter, length 25 mm, purity 99.9%,
GF99322094 Cobalt, foil, thickness 1.0 mm, size 125 × 125 mm, purity 99.9%,