• USA Home
  • Search Results

  • cobaltiiacetatetetrahydrate24908614753111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cobalt(II) acetate tetrahydrate

3 Products
Synonym:   Cobaltous acetate tetrahydrate
Image of Cobalt(II) acetate tetrahydrate
CAS Number:
Linear Formula:
 (CH3COO)2Co · 4H2O
Molecular Weight:
 249.08
 Beilstein/REAXYS Number:
 3731397
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Assay
Show More