• USA Home
  • Search Results

  • cobaltiiifluoride115931002618311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cobalt(III) fluoride

1 Products
Synonym:   Cobalt trifluoride, Cobaltic fluoride
Image of Cobalt(III) fluoride
CAS Number:
Empirical Formula (Hill Notation):
 CoF3
Molecular Weight:
 115.93
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
236136 Cobalt(III) fluoride,