• USA Home
  • Search Results

  • cobaltiinitratehexahydrate291031002622911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cobalt(II) nitrate hexahydrate

4 Products
Synonym:   Cobaltous nitrate hexahydrate, Cobaltous nitrate, Nitric acid, cobalt(II) salt
Image of Cobalt(II) nitrate hexahydrate
CAS Number:
Linear Formula:
 Co(NO3)2 · 6H2O
Molecular Weight:
 291.03
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Special Grade
Show More