• USA Home
  • Search Results

  • cobaltiithiocyanate17510301760511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cobalt(II) thiocyanate

2 Products
Synonym:   Bis(thiocyanic acid)cobalt(II) salt, Cobaltous thiocyanate, Isothiocyanic acid cobalt(II) salt
Image of Cobalt(II) thiocyanate
CAS Number:
Linear Formula:
 Co(SCN)2
Molecular Weight:
 175.10
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More