• USA Home
  • Search Results

  • cocained3solution306376526673111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cocaine-D3 solution

2 Products
Image of Cocaine-D<SUB>3</SUB> solution
CAS Number:
Empirical Formula (Hill Notation):
 C17H18NO4D3
Molecular Weight:
 306.37
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Application(s)
  Package quantity
  Manufacturer designation
  Feature
  Anal. Standard Form
  Special Grade
Show More