• USA Home
  • Search Results

  • codeinesolution299367657311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Codeine solution

2 Products
Image of Codeine solution
CAS Number:
Empirical Formula (Hill Notation):
 C18H21NO3
Molecular Weight:
 299.36
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Application(s)
  Package quantity
  Manufacturer designation
  Feature
  Anal. Standard Form
  Special Grade
Show More