• USA Home
  • Search Results

  • coenzymeb121579581387090111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coenzyme B12

1 Products
Synonym:   5′-Deoxyadenosylcobalamine, Adenosyl cobalamine, Cobamamide, DMBC coenzyme
Image of Coenzyme B<SUB>12</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C72H100CoN18O17P
Molecular Weight:
 1579.58
 Beilstein/REAXYS Number:
 4122932
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
C0884 Coenzyme B12, ≥97.0%,