• USA Home
  • Search Results

  • colchicine399446486811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Colchicine

7 Products
Synonym:   (S)-N-(5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)acetamide
Image of Colchicine
CAS Number:
Empirical Formula (Hill Notation):
 C22H25NO6
Molecular Weight:
 399.44
 Beilstein/REAXYS Number:
 2228813
 EC Number:
 MDL number:
 NACRES:
 NA.72
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Special Grade
  Product Line
Show More