• USA Home
  • Search Results

  • colecalciferol384646797011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Colecalciferol

1 Products
Synonym:   (+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol, Cholecalciferol
Image of Colecalciferol
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O
Molecular Weight:
 384.64
 Beilstein/REAXYS Number:
 2339331
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More