• USA Home
  • Search Results

  • collagenasefromclostridiumhistolyticum12345900112111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Collagenase from Clostridium histolyticum

22 Products
Synonym:   Clostridiopeptidase A
Image of Collagenase from <I>Clostridium histolyticum</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.4.24.3  ( BRENDA | IUBMB )
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Type
  Special Grade
  Ice Shipping
  Purification method
  Specific Activity
  Solubility
  Suitability
  Sterilization
Show More

Product #
Description
C0130 Collagenase from Clostridium histolyticum, for general use, Type I, 0.25-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C5138 Collagenase from Clostridium histolyticum, release of physiologically active rat hepatocytes tested, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C6885 Collagenase from Clostridium histolyticum, release of physiologically active rat epididymal adipocytes tested, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C9891 Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use,
C2674 Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid,
C1639 Collagenase from Clostridium histolyticum, sterile-filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C2139 Collagenase from Clostridium histolyticum, release of rat epididymal adipocytes and hepatocytes tested (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C7657 Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid,
C9263 Collagenase from Clostridium histolyticum, release of physiologically active rat pancreatic islets tested, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid,
C9407 Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture,
C1764 Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat epididymal adipocytes tested, Type II-S, 0.5-5.0 FALGPA units/mg solid,
C8051 Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid,
C9722 Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), 0.5-5.0 FALGPA units/mg solid, cell culture tested,
C5894 Collagenase from Clostridium histolyticum, sterile; sterile-filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C1889 Collagenase from Clostridium histolyticum, sterile-filtered, release of physiologically active rat hepatocytes tested, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid,
C0773 Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, 1,000-3,000 CDU/mg solid (CDU = collagen digestion units),
C9697 Collagenase from Clostridium histolyticum, lyophilized powder (from sterile-filtered solution), Suitable for digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture,
C0255 Collagenase from Clostridium histolyticum, purified by chromatography, ≥1 FALGPA units/mg solid, ≥250 CDU/mg solid (CDU = collagen digestion units), lyophilized powder,
C7926 Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid,
C2799 Collagenase from Clostridium histolyticum, powder, suitable for cell culture, 1000-3000 CDU/mg solid (CDU = collagen digestion units), ≥4 FALGPA units/mg solid, high purity,
C2014 Collagenase from Clostridium histolyticum, sterile-filtered, suitable for release of physiologically active rat pancreatic islets, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid,
C2399 Collagenase from Clostridium histolyticum, sterile-filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, 1,000-3,000 CDU/mg solid,