• USA Home
  • Search Results

  • collagenfromcalfskin12345900734511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Collagen from calf skin

2 Products
CAS Number:
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Package quantity
  Surface coverage
  Sterilization
  Solubility
  Product Line
  Biological Source
  Physical Form
  Suitability
  Restriction
Show More