• USA Home
  • Search Results

  • collagenfromhumanplacenta12345900734511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Collagen from human placenta

7 Products
Synonym:   Collagen Type 4
Image of Collagen from human placenta
CAS Number:
 EC Number:
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Suitability
  Solubility
Show More