• USA Home
  • Search Results

  • congored6966657358011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Congo Red

3 Products
Synonym:   Congo Red 4B, Cosmos Red, Cotton Red B, Cotton Red C, Direct Red 28, Direct Red R, Direct Red Y
Image of Congo Red
CAS Number:
Empirical Formula (Hill Notation):
 C32H22N6Na2O6S2
Molecular Weight:
 696.66
 Beilstein/REAXYS Number:
 3894858
 Colour Index Number:
 22120
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Chemical Composition
  pH Value
  Physical Form
  Special Grade
  Solubility
Show More