• USA Home
  • Search Results

  • copperbis2266tetramethyl35heptanedionate430081404005211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

1 Products
Synonym:   2,2,6,6-Tetramethyl-3,5-heptanedione copper(II) derivative, Cu(TMHD)2
Image of Copper bis(2,2,6,6-<WBR>tetramethyl-<WBR>3,5-<WBR>heptanedionate)
CAS Number:
Linear Formula:
 Cu(OCC(CH3)3CHCOC(CH3)3)2
Molecular Weight:
 430.08
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More