• USA Home
  • Search Results

  • copperiiacetylacetonate261761339516911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper(II) acetylacetonate

3 Products
Synonym:   2,4-Pentanedione copper(II) derivative, Bis(2,4-pentanedionato)copper(II), Cu(acac)2, Cupric acetylacetonate
Image of Copper(II) acetylacetonate
CAS Number:
Linear Formula:
 Cu(C5H7O2)2
Molecular Weight:
 261.76
 Beilstein/REAXYS Number:
 4157957
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Physical Form
Show More