• USA Home
  • Search Results

  • copperiichloride13445744739411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper(II) chloride

5 Products
Synonym:   Cupric chloride
Image of Copper(II) chloride
CAS Number:
Linear Formula:
 CuCl2
Molecular Weight:
 134.45
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
Show More