• USA Home
  • Search Results

  • copperiifluoride10154778919711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper(II) fluoride

1 Products
Synonym:   Cupric fluoride
Image of Copper(II) fluoride
CAS Number:
Empirical Formula (Hill Notation):
 CuF2
Molecular Weight:
 101.54
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
217905 Copper(II) fluoride, 98%,