• USA Home
  • Search Results

  • copperiioxide7955131738011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper(II) oxide

14 Products
Synonym:   Cupric oxide
Image of Copper(II) oxide
CAS Number:
Empirical Formula (Hill Notation):
 CuO
Molecular Weight:
 79.55
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Surface Dimension
  Reaction Suitability
  Particle Dimension
  pH Value
  Potency
  Melting Point
  Density
  Bulk Density
  Special Grade
  Assay
Show More

Product #
Description
544868 Copper(II) oxide, nanopowder, <50 nm particle size (TEM),
450812 Copper(II) oxide, powder, 99.99% trace metals basis,
208841 Copper(II) oxide, powder, <10 μm, 98%,
241741 Copper(II) oxide, ACS reagent, ≥99.0%,
203130 Copper(II) oxide, 99.999% trace metals basis,
792004 Copper(II) oxide, nanorod, diam. × L 10-12 nm × 75-100 nm,
310433 Copper(II) oxide, needles, mixture of CuO and Cu2O, ACS reagent,
1.02764 Copper(II) oxide, granular for elementary analysis,
1.02768 Copper(II) oxide, granular for analysis EMSURE®,
1.02766 Copper(II) oxide, powder for analysis EMSURE® ACS,
1.02761 Copper(II) oxide, powder EMPLURA®,
61200 Copper(II) oxide, puriss. p.a., ≥99.0% (RT), powder,
94737 Copper(II) oxide, ≥99.0% (RT), granular,
450804 Copper(II) oxide, powder, 99.995% trace metals basis,