• USA Home
  • Search Results

  • copperiiphthalocyanine5760714714811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper(II) phthalocyanine

4 Products
Synonym:   CuPc, Phthalocyanine blue, Pigment Blue 15
Image of Copper(II) phthalocyanine
CAS Number:
Empirical Formula (Hill Notation):
 C32H16CuN8
Molecular Weight:
 576.07
 Beilstein/REAXYS Number:
 4121848
 Colour Index Number:
 74160
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  OLED Device Performance
  Special Grade
  Fluorescence Test
  Chemical Composition
  Absorption
  Orbital energy
  OPV Device Performance
Show More