• USA Home
  • Search Results

  • coppersulfate15961775898711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Copper Sulfate

1 Products
Synonym:   Copper(II) sulfate, Cupric sulfate
Image of Copper Sulfate
CAS Number:
Linear Formula:
 CuSO4
Molecular Weight:
 159.61
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More