• USA Home
  • Search Results

  • coproporphyrinitetramethylester710822576720811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coproporphyrin I tetramethyl ester

1 Products
Synonym:   Tetramethyl 3,8,13,18-tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionate
Image of Coproporphyrin I tetramethyl ester
CAS Number:
Empirical Formula (Hill Notation):
 C40H46N4O8
Molecular Weight:
 710.82
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More