• USA Home
  • Search Results

  • corningcostarcellcultureflasks1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Corning® Costar® cell culture flasks

3 Products
Synonym:   cell culture flasks, tissue culture flasks
Image of Corning<SUP>®</SUP> Costar<SUP>®</SUP> cell culture flasks
 eCl@ss
 32040501
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Matrix active group
  Feature
  Manufacturer designation
  Package quantity
  Material
Show More