• USA Home
  • Search Results

  • coronene3003519107111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Coronene

3 Products
Synonym:   Superbenzene, [6]Circulene
Image of Coronene
CAS Number:
Empirical Formula (Hill Notation):
 C24H12
Molecular Weight:
 300.35
 Beilstein/REAXYS Number:
 658468
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More