• USA Home
  • Search Results

  • corticosterone346465022611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Corticosterone

3 Products
Synonym:   11β,21-Dihydroxy-4-pregnene-3,20-dione, 11β,21-Dihydroxyprogesterone, 4-Pregnene-11β,21-diol-3,20-dione, Kendall’s Compound B, Reichstein’s Substance H
Image of Corticosterone
CAS Number:
Empirical Formula (Hill Notation):
 C21H30O4
Molecular Weight:
 346.46
 Beilstein/REAXYS Number:
 2339601
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Biological Source
  Physical Form
Show More