• USA Home
  • Search Results

  • cortisone21acetate402485004411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cortisone 21-acetate

1 Products
Synonym:   17α,21-Dihydroxy-4-pregnene-3,11,20-trione 21-acetate, 21-Acetoxy-4-pregnen-17α-ol-3,11,20-trione, 4-Pregnene-17α,21-diol-3,11,20-trione 21-acetate
Image of Cortisone 21-acetate
CAS Number:
Empirical Formula (Hill Notation):
 C23H30O6
Molecular Weight:
 402.48
 Beilstein/REAXYS Number:
 2067543
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More